(±)-2-Methylbutyric acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
(±)-2-Methylbutanoic acid, 2-Methylbutyric acid, 2-Methylbutanoic acid
A11546.0F
A11546.AC
A11546.AE
A11546.AP
A11546.0FEA
17810
CZK
A11546.0F
A11546.AC
A11546.AE
A11546.AP
(±)-2-Methylbutyric acid ≥98%
(±)-2-Methylbutyric acid
Purity: 98%
CAS No.: 116-53-0
Molecular Formula: C5H10O2
Formula Weight: 102.14
Flash Point: 73°(163°F)
Physical Form: Liquid
Density: 0.937
Refractive Index: 1.4055
Boiling Point: 175-176°
Beilstein Reference: 1720486
UN No.: 3265
MDL No.: MFCD00002669
Vzorec:
C₅H₁₀O₂ MW: 102.13 Bod varu: 175…176 °C Bod tání: –70 °C Hustota: 0.937 Bod vzplanutí: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002669 CAS číslo: 116-53-0 EINECS: 204-145-2 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...