(±)-2-Pentanol, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
(±)-Methyl propyl carbinol, (±)-2-Hydroxy pentane, DL-2-Pentanol, (±)-Propyl methyl carbinol
M-Clarity™ quality level = MQ300
W331600-1KG-K
W331600-4KG-K
SIALW331600-1KG-KEA
3380
CZK
SIALW331600-1KG-K
SIALW331600-4KG-K
(±)-2-Pentanol, Sigma-Aldrich®
(±)-2-Pentanol
Vzorec:
C₅H₁₂O Bod varu: 118 °C (1013 hPa) Bod tání: –50 °C Hustota: 0,81 g/cm³ (20 °C) Bod vzplanutí: 41 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004579 CAS číslo: 6032-29-7 EINECS: 227-907-6 UN: 1105 ADR: 3,III Merck Index: 12,07258 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...