(±)-2-Pentanol, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
(±)-Methyl propyl carbinol, (±)-2-Hydroxy pentane, DL-2-Pentanol, (±)-Propyl methyl carbinol
M-Clarity™ quality level = MQ100
P8017-100ML
P8017-1L
P8017-500ML
SIALP8017-100MLEA
1540
CZK
SIALP8017-100ML
SIALP8017-1L
SIALP8017-500ML
(±)-2-Pentanol, Sigma-Aldrich®
(±)-2-Pentanol
Vzorec:
C₅H₁₂O MW: 88,15 g/mol Bod varu: 118 °C (1013 hPa) Bod tání: –50 °C Hustota: 0,81 g/cm³ (20 °C) Bod vzplanutí: 41 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004579 CAS číslo: 6032-29-7 EINECS: 227-907-6 UN: 1105 ADR: 3,III Merck Index: 12,07258 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...