Tisk…

Bromotrimethylsilane ≥97% stabilizovaný

Dodavatel: Thermo Fisher Scientific
Nebezpečí


Stabilization: Stabilised with copper

A15334.09 A15334.18 A15334.30
A15334.09EA 1430 CZK
A15334.09 A15334.18 A15334.30
Bromotrimethylsilane ≥97% stabilizovaný
Bromotrimethylsilane

Stabilized with copper powder or silver wire


Purity: 97% CAS No.: 2857-97-8 Molecular Formula: C3H9BrSi Formula Weight: 153.09 Storage Temperature: 2°C to 8°C Flash Point: -3°(26°F) Refractive Index: 1.4250 Odour: Acrid Melting Point: -44° Boiling Point: 78-80° Beilstein Reference: 1731135 UN No.: 2924 MDL No.: MFCD00000048

Vzorec: C₃H₉BrSi
Bod varu: 78…80 °C
Bod tání: –44 °C
Hustota: 1,183
Bod vzplanutí: –3 °C (26 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000048
CAS číslo: 2857-97-8
EINECS: 220-672-0
UN: 2924
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR