(±)-2-Heptanol, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(±)-Methyl pentyl carbinol, (±)-2-Hydroxy heptane, Heptan-2-ol, (±)-Pentyl methyl carbinol
M-Clarity™ quality level = MQ100
H3003-100G-A
H3003-25G-A
SIALH3003-100G-AEA
2800
CZK
SIALH3003-100G-A
SIALH3003-25G-A
(±)-2-Heptanol, Sigma-Aldrich®
(±)-2-Heptanol
Vzorec:
C₇H₁₆O MW: 116,2 g/mol Bod varu: 160 °C (1013 hPa) Hustota: 0,82 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004587 CAS číslo: 543-49-7 EINECS: 208-844-3 UN: 1987 ADR: 3,III Merck Index: 13,04682 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...