Tisk…

(±)-3-Amino-1,2-propanediol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

A76001-100G A76001-25G A76001-5G
SIALA76001-100GEA 7850 CZK
SIALA76001-100G SIALA76001-25G SIALA76001-5G
(±)-3-Amino-1,2-propanediol, Sigma-Aldrich®
(±)-3-Amino-1,2-propanediol
Vzorec: C₃H₉NO₂
MW: 91,11 g/mol
Bod varu: 264 °C (1013 hPa)
Hustota: 1,1752 g/cm³ (20 °C)
MDL Number: MFCD00008140
CAS číslo: 616-30-8
EINECS: 210-475-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR