(±)-2-Amino-1-propanol 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
DL-Alaninol, 2-aminopropan-1-ol, DL-2-Amino-1-propanol
L03608.06
L03608.14
L03608.06EA
1460
CZK
L03608.06
L03608.14
(±)-2-Amino-1-propanol 98%
(±)-2-Amino-1-propanol
Purity: 98%
CAS No.: 6168-72-5
Molecular Formula: C3H9NO
Formula Weight: 75.11
Flash Point: 83°(181°F)
Refractive Index: 1.4495
Melting Point: 8°
Boiling Point: 174-176°
Beilstein Reference: 1209234
UN No.: 2735
MDL No.: MFCD00008085
Vzorec:
C₃H₉NO MW: 75,11 g/mol Bod varu: 174…176 °C Bod tání: 8 °C Hustota: 0,965 Bod vzplanutí: 83 °C (181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008085 CAS číslo: 6168-72-5 EINECS: 228-207-3 ADR: 8,III Merck Index: 12,00488 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...