Tisk…

(±)-2-Aminoheptane ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

B22863.14 B22863.22
B22863.14EA 1680 CZK
B22863.14 B22863.22
(±)-2-Aminoheptane ≥98%
(±)-2-Aminoheptane

Purity: 98+% CAS No.: 123-82-0 Molecular Formula: C7H17N Formula Weight: 115.22 Flash Point: 33°(91°F) Refractive Index: 1.4190 Odour: Amine-like pH: 11.45 Boiling Point: 142-144° Beilstein Reference: 635676 Merck Reference: 14,9802 UN No.: 2733 MDL No.: MFCD00008101

Vzorec: C₇H₁₇N
Bod varu: 142…144 °C
Hustota: 0,768
Bod vzplanutí: 33 °C (91 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008101
CAS číslo: 123-82-0
EINECS: 204-655-5
UN: 2733
ADR: 3,III
Merck Index: 14,9802

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR