3-Iodopropionic acid ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
L02655.06
L02655.14
L02655.06EA
851
CZK
L02655.06
L02655.14
3-Iodopropionic acid ≥99%
3-Iodopropionic acid
Purity: 99%
CAS No.: 141-76-4
Molecular Formula: ICH2CH2CO2H
Formula Weight: 199.98
Melting Point: 80-83°
Beilstein Reference: 1699514
UN No.: 3261
MDL No.: MFCD00002765
Vzorec:
C₃H₅IO₂ Bod varu: 260 °C (1013 hPa) Bod tání: 80…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002765 CAS číslo: 141-76-4 EINECS: 205-499-0 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...