(±)-Mercaptosuccinic acid, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
M-Clarity™ quality level = MQ200
M6182-100G
M6182-25G
M6182-500G
SIALM6182-100GEA
3380
CZK
SIALM6182-100G
SIALM6182-25G
SIALM6182-500G
(±)-Mercaptosuccinic acid, Sigma-Aldrich®
(±)-Mercaptosuccinic acid
Vzorec:
C₄H₆O₄S Bod varu: 322 °C (1013 hPa) Bod tání: 150…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004860 CAS číslo: 70-49-5 EINECS: 200-736-4 Merck Index: 12,09479 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...