Lactic acid, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid
M-Clarity™ quality level = MQ200
L6661-100ML
L6661-500ML
SIALL6661-100MLEA
5440
CZK
SIALL6661-100ML
SIALL6661-500ML
Lactic acid, Sigma-Aldrich®
Lactic acid
Vzorec:
H₃CCH(OH)COOH Storage Temperature: Ambient |
MDL Number:
MFCD00004520 CAS číslo: 50-21-5 EINECS: 200-018-0 Merck Index: 13,05351 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...