N-Methylethylenediamine 95% stabilizovaný
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Stabilization:
Stabilised with potassium hydroxide
A12165.06
A12165.14
A12165.22
A12165.06EA
2120
CZK
A12165.06
A12165.14
A12165.22
N-Methylethylenediamine 95% stabilizovaný
N-Methylethylenediamine
Vzorec:
C₃H₁₀N₂ Bod varu: 114…116 °C Hustota: 0,851 Bod vzplanutí: 41 °C (105 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008165 CAS číslo: 109-81-9 EINECS: 203-707-4 UN: 2734 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...