DL-Norleucine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
N1398-100G
N1398-25G
N1398-5G
SIALN1398-100GEA
11910
CZK
SIALN1398-100G
SIALN1398-25G
SIALN1398-5G
DL-Norleucine, Sigma-Aldrich®
DL-Norleucine
Vzorec:
C₆H₁₃NO₂ Bod varu: 246 °C (1013 hPa) Bod tání: 113 °C Hustota: 1,17 g/cm³ (20 °C) |
MDL Number:
MFCD00064422 CAS číslo: 616-06-8 EINECS: 210-462-7 Merck Index: 12,06800 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...