Tisk…

DL-Norleucine ≥98%

Dodavatel: Thermo Fisher Scientific

A10791.14 A10791.22
A10791.14EA 1900 CZK
A10791.14 A10791.22
DL-Norleucine ≥98%
DL-Norleucine

Purity: 98% CAS No.: 616-06-8 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Odour: Odourless Melting Point: 300° Beilstein Reference: 1721748 Merck Reference: 14,6706 MDL No.: MFCD00064422

Vzorec: C₆H₁₃NO₂
Bod varu: 246 °C (1013 hPa)
Bod tání: >300 °C
Hustota: 1,17 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064422
CAS číslo: 616-06-8
EINECS: 210-462-7
Merck Index: 14,6706

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR