1,4-Diiodobutane ≥99% stabilised
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Tetramethylene diiodide
Stabilization:
Stabilised with copper
A16216.14
A16216.22
A16216.14EA
2510
CZK
A16216.14
A16216.22
1,4-Diiodobutane ≥99% stabilised
1,4-Diiodobutane
Stabilized with copper
Purity: 99%
CAS No.: 628-21-7
Molecular Formula: I(CH2)4I
Formula Weight: 309.92
Refractive Index: 1.6212
Melting Point: 5-7°
Boiling Point: 108-110°/10mm
Beilstein Reference: 1098276
MDL No.: MFCD00001099
Vzorec:
C₄H₈I₂ Bod varu: 108…110 °C (10 mmHg) Bod tání: 5…7 °C Hustota: 2,350 Storage Temperature: Ambient |
MDL Number:
MFCD00001099 CAS číslo: 628-21-7 EINECS: 211-032-1 |