3-Chloro-1-iodopropane ≥98% stabilizovaný
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-Chloro-3-iodopropane
Stabilization:
Stabilised with copper
A13489.14
A13489.22
A13489.36
A13489.14EA
2210
CZK
A13489.14
A13489.22
A13489.36
3-Chloro-1-iodopropane ≥98% stabilizovaný
3-Chloro-1-iodopropane
Stabilized with copper
Purity: 98%
CAS No.: 6940-76-7
Molecular Formula: ClCH2CH2CH2I
Formula Weight: 204.44
Flash Point:
Vzorec:
C₃H₆ClI MW: 204,44 g/mol Bod varu: 170…172 °C Hustota: 1,904 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001096 CAS číslo: 6940-76-7 EINECS: 230-088-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...