Tisk…

(±)-2-Chloropropionic acid ≥94%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: α-Chloropropionic Acid, (±)-2-Chloropropanoic acid, 2-Chloropropionic acid

A12860.22 A12860.36
A12860.22EA 872 CZK
A12860.22 A12860.36
(±)-2-Chloropropionic acid ≥94%
(±)-2-Chloropropionic acid

Purity: 94% CAS No.: 598-78-7 Molecular Formula: C3H5ClO2 Formula Weight: 108.53 Flash Point: 101°(213°F) Refractive Index: 1.4350 Melting Point: -13° Boiling Point: 78-80°/11mm Beilstein Reference: 1720259 UN No.: 2511 MDL No.: MFCD00004224

Vzorec: C₃H₅ClO₂
MW: 108,52 g/mol
Bod varu: 78…80 °C (11 mmHg)
Bod tání: –13 °C
Hustota: 1,262
Bod vzplanutí: 101 °C (213 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004224
CAS číslo: 598-78-7
EINECS: 209-952-3
UN: 2511
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR