Tisk…

(±)-3-Bromo-1,2-propanediol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (±)-3-Bromo-1,2-dihydroxy propane, 3-Bromopropane-1,2-diol

M-Clarity™ quality level = MQ200

226130-25G 226130-5G
SIAL226130-25GEA 11750 CZK
SIAL226130-25G SIAL226130-5G
(±)-3-Bromo-1,2-propanediol, Sigma-Aldrich®
(±)-3-Bromo-1,2-propanediol
Vzorec: BrH₂CCH(OH)CH₂OH
MW: 154.99
Bod varu: 72
Hustota: 1.77 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004711
CAS číslo: 4704-77-2
EINECS: 225-186-2
UN: 3265
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR