Tisk…

(±)-1,4-Dibromo-2-butanol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1,4-Dibromo-2-butanol

M-Clarity™ quality level = MQ100

D39185-25G D39185-5G
SIALD39185-25GEA 5890 CZK
SIALD39185-25G SIALD39185-5G
(±)-1,4-Dibromo-2-butanol, Sigma-Aldrich®
(±)-1,4-Dibromo-2-butanol
Vzorec: BrH₂CCH(OH)CH₂CH₂Br
MW: 231,91 g/mol
Bod varu: 114 °C (1013 hPa)
Hustota: 2,023 g/cm³ (0 °C)
MDL Number: MFCD00000252
CAS číslo: 19398-47-1
EINECS: 243-029-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR