(±)-Bromosuccinic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
B81204-100G
B81204-25G
SIALB81204-100GEA
9970
CZK
SIALB81204-100G
SIALB81204-25G
(±)-Bromosuccinic acid, Sigma-Aldrich®
(±)-Bromosuccinic acid
Vzorec:
HOOCBrCHCH₂COOH Bod varu: 319 °C (1013 hPa) Bod tání: 172 °C Hustota: 2,073 g/cm³ (20 °C) |
MDL Number:
MFCD00004213 CAS číslo: 923-06-8 EINECS: 213-087-7 Merck Index: 12,01461 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...