(±)-2-Bromopropionic acid ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
DL-2-Bromopropanoic acid, 2-Bromopropanoic acid, DL-2-Bromopropionic acid, (±)-2-Bromopropanoic acid
A17569.30
A17569.A1
A17569.30EA
1290
CZK
A17569.30
A17569.A1
(±)-2-Bromopropionic acid ≥98%
(±)-2-Bromopropionic acid
Purity: 98%
CAS No.: 598-72-1
Molecular Formula: CH3CHBrCO2H
Formula Weight: 152.98
Flash Point: 100°(212°F)
Refractive Index: 1.4760
Melting Point: 25°
Boiling Point: 202-203°
Beilstein Reference: 605451
UN No.: 3261
MDL No.: MFCD00004211
Vzorec:
H₃CBrCHCOOH MW: 152,98 g/mol Bod varu: 202…203 °C Bod tání: 25 °C Hustota: 1,700 Bod vzplanutí: 100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004211 CAS číslo: 598-72-1 EINECS: 209-947-6 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...