(±)-2-Bromobutyric acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
2-Bromobutanoic acid, (±)-2-Bromobutanoic acid
B21743.0B
B21743.30
B21743.0BEA
4640
CZK
B21743.0B
B21743.30
(±)-2-Bromobutyric acid ≥98%
(±)-2-Bromobutyric acid
Purity: 98%
CAS No.: 80-58-0
Molecular Formula: CH3CH2CHBrCO2H
Formula Weight: 167.01
Flash Point:
Vzorec:
H₃CCH₂BrCHCOOH MW: 167 Bod varu: 99…103 °C (10 mmHg) Bod tání: –4 °C Hustota: 1.56 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004216 CAS číslo: 80-58-0 EINECS: 201-294-5 UN: 3265 Merck Index: 13,01397 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...