L(+)-Histidine monohydrochloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(2S)-(+)-Histidine monohydrochloride, L-3-Imidazol-4-ylalanine monohydrochloride, (S)-(+)-Histidine monohydrochloride, His HCl, (S)-2-Amino-3-(1H-imidazol-4-yl)propanoic acid hydrochloride, H-His-OH·HCl
M-Clarity™ quality level = MQ200
H5659-100G
H5659-1KG
H5659-25G
SIALH5659-100GEA
6860
CZK
SIALH5659-100G
SIALH5659-1KG
SIALH5659-25G
L(+)-Histidine monohydrochloride, Sigma-Aldrich®
L(+)-Histidine monohydrochloride
Vzorec:
C₆H₁₀ClN₃O₂ MW: 191,62 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00064556 CAS číslo: 645-35-2 EINECS: 211-438-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...