Tisk…
Polyvinylpyrrolidone, M.W. ∼ 40,000 (K30), Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP
M-Clarity™ quality level = MQ200
PVP40-100G PVP40-1KG PVP40-500G PVP40-50G PVP40-5KG
SIALPVP40-100GEA 2670 CZK
SIALPVP40-100G SIALPVP40-1KG SIALPVP40-500G SIALPVP40-50G SIALPVP40-5KG
Polyvinylpyrrolidone, M.W. ∼ 40,000 (K30), Sigma-Aldrich®
Polyvinylpyrrolidone
Vzorec: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS číslo: 9003-39-8