Polyvinylpyrrolidone, M.W. ∼ 40,000 (K30), Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Povidone, Polyvinylpyrrolidon, Polyvidone, PVP
M-Clarity™ quality level = MQ200
81420-100G
81420-500G
SIAL81420-100GEA
2290
CZK
SIAL81420-100G
SIAL81420-500G
Polyvinylpyrrolidone, M.W. ∼ 40,000 (K30), Sigma-Aldrich®
Polyvinylpyrrolidone
Vzorec:
(C₆H₉NO)n Storage Temperature: Ambient |
MDL Number:
MFCD01076626 CAS číslo: 9003-39-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...