Polyvinylpyrrolidone, M.W. 44000 - 54000 (av) Ph. Eur. (K30)
Dodavatel: VWR Chemicals
Synonyms:
Povidone, Polyvinylpyrrolidon, Polyvidone, PVP
26617.291EA
16780
CZK
26617.291
26617.360
Polyvinylpyrrolidone, M.W. 44000 - 54000 (av) Ph. Eur. (K30)
Polyvinylpyrrolidone
Vzorec:
(C₆H₉NO)n Storage Temperature: Ambient |
MDL Number:
MFCD01076626 CAS číslo: 9003-39-8 |
Výsledky specifikačního testu
Assay (N) (calculated on anhydrous) | 11.5 - 12.8 % |
Appearance | Conforms (see CoA/CoS) |
Identification A | Passes test |
Identification E | Passes test |
Solution S1 | Passes test |
Appearance of solution | Passes test |
pH (5 %) | 3.0 - 5.0 |
Viscosity (expressed as K-value) | 27.0 - 32.4 |
Aldehydes (as C2H4O) | Max. 500 ppm |
Peroxides (as H2O2) | Max. 400 ppm |
Formic acid | Max. 0.5 % |
Hydrazine | Max. 1 ppm |
Impurity A | Max. 10 ppm |
Impurity B | Max. 3.0 % |
Heavy metals (as Pb) | Max. 10 ppm |
Water | Max. 5.0 % |
Sulphated ash | Max. 0.1 % |
Residual solvents | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...