Tisk…
Polyvinylpyrrolidone, M.W. ∼ 10,000 (K15), Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP
M-Clarity™ quality level = MQ200
PVP10-100G PVP10-500G PVP10-12KG PVP10-1KG
SIALPVP10-100GEA 2160 CZK
SIALPVP10-100G SIALPVP10-500G SIALPVP10-12KG SIALPVP10-1KG
Polyvinylpyrrolidone, M.W. ∼ 10,000 (K15), Sigma-Aldrich®
Polyvinylpyrrolidone
Vzorec: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS číslo: 9003-39-8