Tanin ACS
Dodavatel: Thermo Fisher Scientific
Varování
|
Synonyms:
Gallotannin, Tannin
36410.22
36410.36
AB208064.0100
36410.22EA
1610
CZK
36410.22
36410.36
ABCRAB208064.0100
Tanin ACS
Tanin
CAS No.: 1401-55-4
Molecular Formula: C76H52O46
Formula Weight: 1701.23
Flash Point: 199°(390°F)
Physical Form: Powder
Sensitivity: Air and Light Sensitive
pH: 3.5
Melting Point: ca 218° dec.
Solubility: Soluble in water, warm glycerol, alcohol, acetone
Beilstein Reference: 8186396
Merck Reference: 14,9052
MDL No.: MFCD00066397
Vzorec:
C₇₆H₅₂O₄₆ MW: 1701.2 g/mol Bod tání: ∼ 218 °C (rozkl.) Hustota: Bod vzplanutí: 199 °C (390 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00066397 CAS číslo: 1401-55-4 EINECS: 215-753-2 Merck Index: 14,9052 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...