Tisk…

L(+)-vinan disodný dihydrát, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Sodium L-(+)-tartrate dihydrate, L(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Sodium tartrate dibasic dihydrate, Disodium L-(+)-Tartrate Dihydrate, (R,R)-(+)-Disodium tartrate dihydrate, L(+)-Tartaric acid, disodium salt, dihydrate, Sodium L-tartrate dibasic dihydrate, L(+)-Sodium tartrate dibasic dihydrate, Sodium Tartrate, Dihydrate, (+)-Disodium L-tartrate dihydrate, (+)-Disodium-L-tartrat dihydrate

M-Clarity™ quality level = MQ200

228729-5G 228729-100G 228729-500G
SIAL228729-5GEA 1180 CZK
SIAL228729-5G SIAL228729-100G SIAL228729-500G
L(+)-vinan disodný dihydrát, Sigma-Aldrich®
L(+)-vinan disodný dihydrát
Vzorec: NaOOC(CHOH)₂COONa·2H₂O
MW: 230,08 g/mol
Bod tání: 57 °C
Hustota: 1,818 g/cm³ (20 °C)
MDL Number: MFCD00150035
CAS číslo: 6106-24-7
EINECS: 212-773-3
Merck Index: 12,08833
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39a75038f4ff5e6c234267b15540823ae361bed4&vid=89289572c48b0c6415f8f11e916a51f66e129474&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR