L(+)-Asparagin monohydrát, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
L-Asparagine monohydrate, H-Asn-OH·H₂O, (S)-(+)-Asparagine monohydrate, (S)-2,4-Diamino-4-oxobutanoic acid hydrate, H-Asn-OH.H2O
M-Clarity™ quality level = MQ200
A8381-100G
A8381-1KG
SIALA8381-100GEA
3620
CZK
SIALA8381-100G
SIALA8381-1KG
L(+)-Asparagin monohydrát, Sigma-Aldrich®
L(+)-Asparagine monohydrate
Vzorec:
C₄H₈N₂O₃·1H₂O MW: 150,13 g/mol Bod tání: 215…217 °C Hustota: 1,567 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00151038 CAS číslo: 5794-13-8 EINECS: 200-735-9 Merck Index: 14,00837 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...