Tisk…

L(+)-Asparagin monohydrát ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: L-Asparagine monohydrate, H-Asn-OH·H₂O, (S)-(+)-Asparagine monohydrate, (S)-2,4-Diamino-4-oxobutanoic acid hydrate, H-Asn-OH.H2O

A15012.0E A15012.22 A15012.36
A15012.0EEA 21250 CZK
A15012.0E A15012.22 A15012.36
L(+)-Asparagin monohydrát ≥98%
L(+)-Asparagin monohydrát

Purity: 98+% CAS No.: 5794-13-8 Molecular Formula: C4H8N2O3.H2O Formula Weight: 150.14 (132.12anhy) Melting Point: ca 245° dec. Beilstein Reference: 5767869 Merck Reference: 14,837 Optical Rotation: +31° (c=5 in 5N HCl) MDL No.: MFCD00151038

Vzorec: C₄H₈N₂O₃·1H₂O
Hustota: 1,543
Storage Temperature: Ambient
MDL Number: MFCD00151038
CAS číslo: 5794-13-8
EINECS: 200-735-9
Merck Index: 14,837

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR