TRITON™ X-100
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol
CAS No.: 9002-93-1
Formula Weight: ca. 625
Flash Point: 180°(356°F)
Physical Form: Liquid
Density: 1.061
Refractive Index: 1.4900
Odour: Mild
Melting Point: ca 5°
Boiling Point:
Detergent, equivalent to Nonidet P-40
Bod varu:
>250 °C Bod tání: ∼ 5 °C Hustota: 1.061 Bod vzplanutí: 180 °C (356 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00132505 CAS číslo: 9002-93-1 UN: 3082 ADR: 9 III Merck Index: 14,6761 |