Polyethylene glycol 35,000, Sigma-Aldrich®
Dodavatel: Merck
94646-1KG-F
94646-250G-F
SIAL94646-1KG-FEA
4110
CZK
SIAL94646-1KG-F
SIAL94646-250G-F
Polyethylene glycol 35,000, Sigma-Aldrich®
Polyethylene glycol 35,000
Vzorec:
H(OCH₂CH₂)nOH Storage Temperature: Ambient |
MDL Number:
MFCD00081839 CAS číslo: 25322-68-3 EINECS: 500-038-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...