Polyethylenglykol 3000, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
81227-1KG
81227-2.5KG
81227-250G
SIAL81227-1KGEA
4730
CZK
SIAL81227-1KG
SIAL81227-2.5KG
SIAL81227-250G
Polyethylenglykol 3000, Sigma-Aldrich®
Polyethylenglykol 3000
Vzorec:
H(OCH₂CH₂)nOH Bod tání: 55…58 °C Hustota: 1,21 g/cm³ (20 °C) Storage Temperature: Ambient |
CAS číslo:
25322-68-3 EINECS: 500-038-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...