Platinum rhodium alloy (87:13 w%), wire for ISA type R standard grade thermocouple, Ø 0.127 mm (0.005 in)
Dodavatel: Thermo Fisher Scientific
42904.G5
42904.G9
42904.G5EA
27110
CZK
42904.G5
42904.G9
Platinum rhodium alloy (87:13 w%), wire for ISA type R standard grade thermocouple, Ø 0.127 mm (0.005 in)
Platinum rhodium alloy (87:13 w%)
Molecular Formula: Pt:Rh; 87:13 wt%
Physical Form: ≈0.25g/m
Odour: Odourless
MDL No.: MFCD00198186