Nickel chromium alloy (80:20 w%), wire, Ø 0.2 mm (0.008 in)
Dodavatel: Thermo Scientific
|
Varování
|
45057.H4
45057.G9
45057.H4EA
1930
CZK
45057.H4
45057.G9
Nickel chromium alloy (80:20 w%), wire, Ø 0.2 mm (0.008 in)
Nickel chromium alloy (80:20 w%)
CAS No.: 11106-97-1
Molecular Formula: Ni:Cr; 80:20 wt%
Physical Form: ≈0.21g/m
Odour: Odourless
MDL No.: MFCD00673110
Storage Temperature:
Ambient |
MDL Number:
MFCD00673110 CAS číslo: 11106-97-1 |