Nickel chromium alloy (80:20 w%), wire, Ø 0.51 mm (0.02 in)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
13967.H4
13967.H9
13967.H4EA
3920
CZK
13967.H4
13967.H9
Nickel chromium alloy (80:20 w%), wire, Ø 0.51 mm (0.02 in)
Nickel chromium alloy (80:20 w%)
CAS No.: 11106-97-1
Molecular Formula: Ni:Cr; 80:20 wt%
Physical Form: ≈1.86g/m
Odour: Odourless
MDL No.: MFCD00673110
Storage Temperature:
Ambient |
MDL Number:
MFCD00673110 CAS číslo: 11106-97-1 |