Tisk…
Copper nickel alloy (55:45 w%), wire, Ø 0.13 mm (0.005 in)
Dodavatel: Thermo Fisher Scientific
Nebezpečí
43270.H4 43270.H9
43270.H4EA 1280 CZK
43270.H4 43270.H9
Copper nickel alloy (55:45 w%), wire, Ø 0.13 mm (0.005 in)
Copper nickel alloy (55:45 w%)
Molecular Formula: Cu:Ni 55:45 wt% Physical Form: ≈0.11g/m Odour: Odourless MDL No.: MFCD00801098