(±)-3-Butyne-2-ol 55% ∼7.5 mol/l in aqueous solution for synthesis, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
(±)-But-3-yn-2-ol, (±)-3-Hydroxy-1-butyne
M-Clarity™ quality level = MQ200
8.20230.0100
8.20230.0500
8.20230.0100EA
1110
CZK
8.20230.0100
8.20230.0500
(±)-3-Butyne-2-ol 55% ∼7.5 mol/l in aqueous solution for synthesis, Sigma-Aldrich®
(±)-3-Butyne-2-ol
Vzorec:
C₄H₆O MW: 70.09 Bod tání: -12 Hustota: 0.95 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004541 CAS číslo: 2028-63-9 EINECS: 217-978-1 UN: 2929 ADR: 6.1 (3) II |