Guma tragacanth, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Tragacanth, Tragacanth gum
M-Clarity™ quality level = MQ200
G1128-100G
G1128-1KG
G1128-500G
SIALG1128-100GEA
4270
CZK
SIALG1128-100G
SIALG1128-1KG
SIALG1128-500G
Guma tragacanth, Sigma-Aldrich®
Guma tragacanth
Storage Temperature:
Ambient |
MDL Number:
MFCD00131255 CAS číslo: 9000-65-1 EINECS: 232-552-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...