Guma tragacanth, powder
Dodavatel: Thermo Scientific
Synonyms:
Tragacanth, Tragacanth gum
A18502.22
A18502.36
A18502.22EA
2250
CZK
A18502.22
A18502.36
Guma tragacanth, powder
Guma tragacanth
CAS No.: 9000-65-1
Odour: Odourless
Merck Reference: 14,4581
MDL No.: MFCD00131255
Storage Temperature:
Ambient |
MDL Number:
MFCD00131255 CAS číslo: 9000-65-1 EINECS: 232-552-5 Merck Index: 14,4581 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...