Tisk…

Starch (from wheat), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

S5127-100G S5127-500G
SIALS5127-100GEA 967 CZK
SIALS5127-100G SIALS5127-500G
Starch (from wheat), Sigma-Aldrich®
Starch
Vzorec: (C₆H₁₀O₅)n
MW: 162.14
Hustota: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS číslo: 9005-25-8
EINECS: 232-679-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR