Tisk…

Agaróza, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

05066-500G 05066-50G
SIAL05066-500GEA 60560 CZK
SIAL05066-500G SIAL05066-50G
Agaróza, Sigma-Aldrich®
Agaróza
Bod tání: 60…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS číslo: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR