Agar, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Agar Agar
MQ200
A6686-100G
A6686-1KG
A6686-500G
SIALA6686-100GEA
4320
CZK
SIALA6686-100G
SIALA6686-1KG
SIALA6686-500G
Agar, Sigma-Aldrich®
Agar
Vzorec:
C₁₂nH₁₈nOn₉n Bod tání: 90 Storage Temperature: Ambient |
MDL Number:
MFCD00081288 CAS číslo: 9002-18-0 EINECS: 232-658-1 Merck Index: 13,00184 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...