Tisk…

Agar, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Agar Agar

MQ200

A4550-1KG A4550-500G A4550-5KG
SIALA4550-1KGEA 13950 CZK
SIALA4550-1KG SIALA4550-500G SIALA4550-5KG
Agar, Sigma-Aldrich®
Agar
Vzorec: C₁₂nH₁₈nOn₉n
Bod tání: 90
Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS číslo: 9002-18-0
EINECS: 232-658-1
Merck Index: 13,00184
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR