Tisk…

Agar plant cell culture tested

Dodavatel: Thermo Fisher Scientific

Synonyms: Agar Agar

H26724.0E H26724.22 H26724.36
H26724.0EEA 38850 CZK
H26724.0E H26724.22 H26724.36
Agar plant cell culture tested
Agar

CAS No.: 9002-18-0 Odour: Odourless Merck Reference: 14,184 MDL No.: MFCD00081288

Bod tání: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS číslo: 9002-18-0
EINECS: 232-658-1
Merck Index: 14,184
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR