Tisk…

Neomycin trisulphate salt hydrate, powder, BioReagent cell culture reagent, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ200

N6386-100G N6386-25G N6386-5G
SIALN6386-100GEA 11720 CZK
SIALN6386-100G SIALN6386-25G SIALN6386-5G
Neomycin trisulphate salt hydrate, powder, BioReagent cell culture reagent, Sigma-Aldrich®
Neomycin trisulphate salt hydrate
Vzorec: C₂₃H₄₆N₆O₁₃·₃H₂SO₄·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00150829
CAS číslo: 1405-10-3
EINECS: 215-773-1
Merck Index: 12,06542

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR