Neomycin trisulphate salt hydrate, powder, meets analytical specification of USP, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Neomycin B, Antibiotic 10676, Neomycin trisulfate salt hydrate, Neomycin trisulphate salt hydrate
M-Clarity™ quality level = MQ200
N5285-100G
N5285-25G
SIALN5285-100GEA
21840
CZK
SIALN5285-100G
SIALN5285-25G
Neomycin trisulphate salt hydrate, powder, meets analytical specification of USP, Sigma-Aldrich®
Neomycin trisulphate salt hydrate
Vzorec:
C₂₃H₄₆N₆O₁₃·₃H₂SO₄·xH₂O MW: 908.88 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00150829 CAS číslo: 1405-10-3 EINECS: 215-773-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...