Tisk…

Gentamicin sulphate, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ200

G4918-250MG G4918-5G
SIALG4918-250MGEA 6000 CZK
SIALG4918-250MG SIALG4918-5G
Gentamicin sulphate, Sigma-Aldrich®
Gentamicin sulphate
Bod tání: 218…237 °C
Storage Temperature: Ambient
MDL Number: MFCD00270181
CAS číslo: 1405-41-0
EINECS: 215-778-9
Merck Index: 13,04403

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR