Tisk…
Gentamicin sulphate, Sigma-Aldrich®
Dodavatel: Merck
Nebezpečí
Synonyms: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
M-Clarity™ quality level = MQ300
G3632-1G G3632-10G G3632-250MG G3632-25G G3632-5G
SIALG3632-1GEA 4670 CZK
SIALG3632-1G SIALG3632-10G SIALG3632-250MG SIALG3632-25G SIALG3632-5G
Gentamicin sulphate, Sigma-Aldrich®
Gentamicin sulphate
Bod tání: 218…237 °C
Storage Temperature: Ambient
MDL Number: MFCD00270181
CAS číslo: 1405-41-0
EINECS: 215-778-9
Merck Index: 13,04403